missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIAPH-2 Antibody (CL1111), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52966
This item is not returnable.
View return policy
Description
DIAPH-2 Monoclonal antibody specifically detects DIAPH-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Specifications
| DIAPH-2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| CL1111 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated | |
| DIA2, DIAdiaphanous (Drosophila, homolog) 2, diaphanous homolog 2 (Drosophila), Diaphanous-related formin-2, diaphorase-2, DRF2, POF, POF2FLJ11167, protein diaphanous homolog 2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GGSEEPGGGRSNKRSAGNRAANEEETKNKPKLNIQIKTLADDVRDRITSFRKSTVKKEKPLIQHPIDSQVAMSEFPAAQPLYDERSLNLSEKEVLDLFEKMMEDMNLNEEKKAPLRNKDFTTKREMVVQYISAT | |
| 0.1 mL | |
| Cell Cycle and Replication, Transcription Factors and Regulators | |
| 1730 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction