missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIAPH-2 Antibody (CL1111) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52966IR
This item is not returnable.
View return policy
Description
DIAPH-2 Monoclonal antibody specifically detects DIAPH-2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| DIAPH-2 | |
| Monoclonal | |
| DyLight 755 | |
| DIA2, DIAdiaphanous (Drosophila, homolog) 2, diaphanous homolog 2 (Drosophila), Diaphanous-related formin-2, diaphorase-2, DRF2, POF, POF2FLJ11167, protein diaphanous homolog 2 | |
| This antibody was developed using a recombinant protein derived from O60879, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Cycle and Replication, Transcription Factors and Regulators | |
| 1730 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL1111 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction