missing translation for 'onlineSavingsMsg'
Learn More

Diamylamine, TRC

Product Code. 30382170
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382170 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382170 Supplier Toronto Research Chemicals Supplier No. TRCD4940955G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diamylamine
CAS 2050-92-2
Molecular Formula C10H23N
Quantity 5 g
Synonym Dipentylamine, N-Pentyl-1-pentanamine, Di-n-amylamine, Di-n-pentylamine, Diamylamine, Dipentanamine, N,N-Dipentylamine, NSC 6329
InChI Formula InChI=1S/C10H23N/c1-3-5-7-9-11-10-8-6-4-2/h11H,3-10H2,1-2H3
SMILES CCCCCNCCCCC
Molecular Weight (g/mol) 157.3
Formula Weight 157.183
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.