missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diamond powder, synthetic, 40-60 micron, 99.9% (metals basis)

Diamond powder is generally used as an abrasive to grind and polish hard surfaces

Brand:  Alfa Aesar™ 013402.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11376718

  • £323.30 / 5g


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Specifications

Chemical Identifiers

Diamond powder
Synthetic
MFCD00211867

Specifications

Diamond powder
Synthetic
Powder
(metals basis)
5g
40 to 60 Micron
99.9%
MFCD00211867
Videos
SDS