missing translation for 'onlineSavingsMsg'
Learn More

Diammonium-2,2’ Azino-bis(3-ethylbenzothiazoline-6-sulfonate), TRC

Product Code. 30136318
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136318 5 g 5g
30134043 25 g 25g
17970685 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136318 Supplier Toronto Research Chemicals Supplier No. TRCD4167755G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30931-67-0
Molecular Formula C18 H18 N4 O6 S4 . 2 H3 N
Molecular Weight (g/mol) 548.68
Synonym 2,2'-(1,2-Hydrazinediylidene)bis[3-ethyl-2,3-dihydro-6-benzothiazolesulfonic Acid Ammonium Salt (1:2), 2,2'-Azinobis[3-ethyl-2,3-dihydro-6-benzothiazolesulfonic Acid Diammonium Salt, 3-Ethyl-2-oxo-6-Benzothiazolinesulfonic Acid Azine Diammonium Salt, 2,2'-Azino-bis-(3-ethyl-benzthiazoline-6-sulfonate) Diammonium Salt, 2,2'-Azino-bis-(3-ethylbenzothazoline-6-sulfonate), 2,2'-Azinobis(3-ethylbenzothiazoline-6-sulfonic acid) Diammonium Salt, ABTS Diammonium Salt, Diammonium 2,2'-Azinobis(3-ethyl-6-benzothiazolinesulfonate)
IUPAC Name azane;(2Z)-3-ethyl-2-[(Z)-(3-ethyl-6-sulfo-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonic acid
SMILES N.N.CCN1\C(=N\N=C\2/Sc3cc(ccc3N2CC)S(=O)(=O)O)\Sc4cc(ccc14)S(=O)(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C18H18N4O6S4.2H3N/c1-3-21-13-7-5-11(31(23,24)25)9-15(13)29-17(21)19-20-18-22(4-2)14-8-6-12(32(26,27)28)10-16(14)30-18;;/h5-10H,3-4H2,1-2H3,(H,23,24,25)(H,26,27,28);2*1H3/b19-17-,20-18-;;
Formula Weight 548.064
Chemical Name or Material Diammonium-2,2' Azino-bis(3-ethylbenzothiazoline-6-sulfonate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.