missing translation for 'onlineSavingsMsg'
Learn More

Diaminomaleonitrile, 98%

Product Code. 10341034 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10341034 500 g Glass bottle 500g
10614672 25 g Glass bottle 25g
10104290 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10341034 Supplier Thermo Scientific Acros Supplier No. 172705000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1187-42-4 | C4H4N4 | 108.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1187-42-4
Molecular Formula C4H4N4
Molecular Weight (g/mol) 108.1
MDL Number MFCD00001870
InChI Key DPZSNGJNFHWQDC-ARJAWSKDSA-N
Synonym diaminomaleonitrile, 2,3-diaminomaleonitrile, hydrogen cyanide tetramer, maleonitrile, diamino, ccris 921, 2-butenedinitrile, 2,3-diamino-, 2z, 2z-diaminobut-2-enedinitrile, 2,3-diamino maleonitrile, 2-butenedinitrile, 2,3-diamino-, z, z-2,3-diaminobut-2-enedinitrile
PubChem CID 2723951
IUPAC Name (Z)-2,3-diaminobut-2-enedinitrile
SMILES C(#N)C(=C(C#N)N)N

Specifications

Melting Point 178.0°C to 179.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NCC(NH2)=C(NH2)CN
Quantity 500 g
Beilstein 04, II, 949
Fieser 05,175; 04,480; 13,327; 01,1256; 02,454; 04,565; 06,657; 07,414; 08,524; 09,512; 10,455; 13,301
Solubility Information Solubility in water: 5g/L (25°C). Other solubilities: soluble in methanol, ethanol, dmf and dimethyl, sulfoxide, slightly soluble in ethyl ether and dioxane, insol. in benzene,acetone,xylene,toluene and cs2
Formula Weight 108.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Diaminomaleonitrile
Show More Show Less
compliance-icons
Product Identifier
  • Diaminomaleonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.