missing translation for 'onlineSavingsMsg'
Learn More

Diallyldimethylammonium Chloride (60% w/w in Water), TRC

Product Code. 30319573
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319573 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319573 Supplier Toronto Research Chemicals Supplier No. TRCA559015100ML
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diallyldimethylammonium Chloride
CAS 7398-69-8
Molecular Formula C8H16ClN
Quantity 100 mL
Synonym N-Allyl-N,N-dimethylprop-2-en-1-aminium Chloride, AG Fix M 80, C 1615, C 1615M-BR 30, D 1259, DADMAC, Dimethyldiallylammonium Chloride, Hercules D 1259, LYFM 205-65, M-DADMAC, N,N-Diallyl-N,N-dimethylammonium Chloride, Nalco 7544, Nalco TX 13645, Newfrontier C 1615
InChI Formula InChI=1S/C8H16N.ClH/c1-5-7-9(3,4)8-6-2;/h5-6H,1-2,7-8H2,3-4H3;1H/q+1;/p-1
SMILES [Cl-].C[N+](C)(CC=C)CC=C
IUPAC Name dimethyl-bis(prop-2-enyl)azanium;chloride
Molecular Weight (g/mol) 161.67
Formula Weight 161.0971
Concentration or Composition (by Analyte or Components) 60 %
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.