missing translation for 'onlineSavingsMsg'
Learn More

Diallylamine, 97%

Product Code. 10510551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10510551 250 mL 250mL
15760789 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10510551 Supplier Thermo Scientific Acros Supplier No. 112042500
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 124-02-7 | C6H11N | 97.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-02-7
Molecular Formula C6H11N
Molecular Weight (g/mol) 97.16
MDL Number MFCD00008642
InChI Key DYUWTXWIYMHBQS-UHFFFAOYSA-N
Synonym diallylamine, di-2-propenylamine, 2-propen-1-amine, n-2-propenyl, amine, diallyl, bis prop-2-en-1-yl amine, n-2-propenyl-2-propen-1-amine, unii-n18exb6v6p, n,n-diallylamine, ccris 4776, n-allylprop-2-en-1-amine
PubChem CID 31279
IUPAC Name N-prop-2-enylprop-2-en-1-amine
SMILES C=CCNCC=C

Specifications

Melting Point -88.0°C
Color Colorless to Yellow
Density 0.7890g/mL
Boiling Point 110.0°C to 112.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (H2C=CHCH2)2NH
Refractive Index 1.4395 to 1.4415
Quantity 250 mL
Beilstein 04, 208
Merck Index 15, 2974
Specific Gravity 0.789
Solubility Information Solubility in water: 90g/L in water. Other solubilities: soluble in diethyl ether, ethanol
Formula Weight 97.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diallylamine, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diallylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.