missing translation for 'onlineSavingsMsg'
Learn More

Diallyl Phthalate, TRC

Product Code. 30364705
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30364705

Brand: Toronto Research Chemicals TRCD4160551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-allyl ester
CAS 131-17-9
Molecular Formula C14 H14 O4
Quantity 1 g
Synonym 1,2-Benzenedicarboxylic acid, 1,2-di-2-propen-1-yl ester (ACI), 1,2-Benzenedicarboxylic acid, di-2-propenyl ester (9CI), Phthalic acid, diallyl ester (6CI, 8CI), Allyl phthalate, Daiso DAP monomer, DAP, DAP Monomer, Dap Tohto DT 170, DAP-M, Dapon R, Dappu, DCP-A, Diallyl phthalate, DT 170, KY 5200, Monoplex DAP, NSC 7667
InChI Formula InChI=1S/C14H14O4/c1-3-9-17-13(15)11-7-5-6-8-12(11)14(16)18-10-4-2/h3-8H,1-2,9-10H2
SMILES C=CCOC(=O)c1ccccc1C(=O)OCC=C
IUPAC Name bis(prop-2-enyl) benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 246.26
Formula Weight 246.0892
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.