missing translation for 'onlineSavingsMsg'
Learn More

Diallyl phthalate, 98%

Product Code. 10434935 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10434935 2.5 L Glass bottle 2.5 L
10559453 250 mL Glass bottle 250mL
10741721 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10434935 Supplier Thermo Scientific Acros Supplier No. 276620025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 131-17-9 | C14H14O4 | 246.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-17-9
Molecular Formula C14H14O4
Molecular Weight (g/mol) 246.26
MDL Number MFCD00008646
InChI Key QUDWYFHPNIMBFC-UHFFFAOYSA-N
Synonym diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st
PubChem CID 8560
IUPAC Name bis(prop-2-enyl) benzene-1,2-dicarboxylate
SMILES C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C

Specifications

Melting Point -70.0°C
Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 300.0°C
Flash Point 166°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5180 to 1.5200
Quantity 2.5 L
UN Number 3082
Beilstein 09, 4120
Specific Gravity 1.12
Solubility Information Solubility in water: 148mg/l (20°C)
Viscosity 13 mPa.s (20°C)
Formula Weight 246.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diallyl phthalate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diallyl phthalate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P273-Avoid release to the environment.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.