missing translation for 'onlineSavingsMsg'
Learn More

Diallyl phthalate, 97%

Product Code. 11480508
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480508 100 g 100g
11490508 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480508 Supplier Thermo Scientific Alfa Aesar Supplier No. B24648.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 131-17-9 | C14H14O4 | 246.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-17-9
Molecular Formula C14H14O4
Molecular Weight (g/mol) 246.262
MDL Number MFCD00008646
InChI Key QUDWYFHPNIMBFC-UHFFFAOYSA-N
Synonym diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st
PubChem CID 8560
IUPAC Name bis(prop-2-enyl) benzene-1,2-dicarboxylate
SMILES C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C

Specifications

Melting Point -70°C
Density 1.12
Boiling Point 160°C to 163°C (4 mmHg)
Flash Point 166°C (330°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.519
Quantity 100 g
UN Number UN3082
Beilstein 1880877
Formula Weight 246.27
Percent Purity 97%
Chemical Name or Material Diallyl phthalate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.