missing translation for 'onlineSavingsMsg'
Learn More

Diallyl Disulphide (Technical grade, >80%), TRC

Product Code. 30354672
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
Product Code. Quantity unitSize
30354672 1 g 1g
30363678 100 g 100g
30364878 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 30354672

Brand: Toronto Research Chemicals TRCD4160351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2179-57-9
Formula Weight 146.0224
InChI Formula InChI=1S/C6H10S2/c1-3-5-7-8-6-4-2/h3-4H,1-2,5-6H2
IUPAC Name 3-(prop-2-enyldisulfanyl)prop-1-ene
Molecular Formula C6 H10 S2
Molecular Weight (g/mol) 146.27
SMILES C=CCSSCC=C
Synonym Diallyl Disulphide, Allyl Disulfide
Chemical Name or Material Allyl Disulfide

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.