missing translation for 'onlineSavingsMsg'
Learn More

Diallyl disulfide, tech. 80%, remainder mainly diallyl sulfide and diallyl trisulfide, Thermo Scientific Chemicals

Catalog Number 11377556
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2179-57-9
C6H10S2
146.27
MFCD00008656
PFRGXCVKLLPLIP-UHFFFAOYSA-N
diallyl disulfide, allyl disulfide, diallyldisulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 2-propenyl disulphide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene
16590
CHEBI:4488
3-(prop-2-enyldisulfanyl)prop-1-ene
C=CCSSCC=C
This item is not returnable. View return policy
2179-57-9
C6H10S2
146.27
MFCD00008656
PFRGXCVKLLPLIP-UHFFFAOYSA-N
diallyl disulfide, allyl disulfide, diallyldisulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 2-propenyl disulphide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene
16590
CHEBI:4488
3-(prop-2-enyldisulfanyl)prop-1-ene
C=CCSSCC=C

Diallyl disulfide, 80%, C6H10S2, CAS Number-2179-57-9, diallyldisulfide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene, 2-propenyl disulphide, diallyl disulfide, allyl disulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 100g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 180°C to 190°C
Flash Point 51°C (123°F)
Odor Garlic-like
Linear Formula CH2=CHCH2S-SCH2CH=CH2
Quantity 5 g
UN Number UN1993
Beilstein 1699241
Formula Weight 146.27
Percent Purity 80%
Grade Technical
Assay remainder mainly diallyl sulfide and diallyl trisulfide
Chemical Name or Material Diallyl disulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'