missing translation for 'onlineSavingsMsg'
Learn More

Diaion™ WA30, ion exchange resin, weakly basic porous type, 1.5 meq/ml on poly(styrene-divinylbenzene)

Product Code. 15415025
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415025 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415025 Supplier Thermo Scientific Alfa Aesar Supplier No. 046592.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65405-58-5 | C21H25N | 291.438 g/mol

Water treatment, refining of starch syrup and glucose, refining of beet sugar

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form 300-1180μm Beads
CAS 65405-58-5
Odor Odorless
Particle Size 300 to 1180μm
Molecular Formula C21H25N
Color Yellow
Synonym divinylbenzene, vinylbenzyl dimethylamine polymer, benzenemethanamine, ar-ethenyl-n,n-dimethyl-, polymer with diethenylbenzene, 2-ethenylphenyl methyl dimethylamine; divinylbenzene, 1,2-bis ethenyl benzene; 1-2-ethenylphenyl-n,n-dimethylmethanamine
Quantity 1 kg
InChI Key OTCJDTFYONDHFM-UHFFFAOYSA-N
SMILES CN(C)CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1C=C
IUPAC Name 1,2-bis(ethenyl)benzene;1-(2-ethenylphenyl)-N,N-dimethylmethanamine
Molecular Weight (g/mol) 291.438
PubChem CID 6455131
Chemical Name or Material Diaion™ WA30, ion exchange resin, Weakly basic porous type, 1.5meq/mL on poly(styrene-divinylbenzene)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.