Learn More
Diaion™ UBK12(Na), ion exchange resin, 12% cross-linked, strongly acidic gel type, 2.3 meq/ml on PS-DVB, Thermo Scientific™
Softening, metal recovery, amino acid separation, sugar refining
Brand: Thermo Scientific Alfa Aesar 046671.A1
Code : NEW
Additional Details : Weight : 1.00000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
600-700μm Beads | |
12% cross-linked, strongly acidic gel type, 2.3meq/mL on PS-DVB | |
1 kg |
Diaion™ UBK12 (Na), ion exchange resin | |
600 to 700μm |