Learn More
Diaion™ UBA100(Cl), ion exchange resin, cross-linked, strongly basic gel type, 1.3 meq/ml on PS-DVB, Thermo Scientific™
Power industry condensate demineralization, metal recovery, catalysis
Brand: Thermo Scientific Alfa Aesar 046938.A1
Code : NEW
Additional Details : Weight : 1.00000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
500-600μm Beads | |
Cross-linked, strongly basic gel type, 1.3meq/mL on PS-DVB | |
1 kg |
Diaion™ UBA100 (Cl), ion exchange resin | |
500 to 600μm |