Learn More
Thermo Scientific Alfa Aesar Diaion™ SK110(Na), ion exchange resin, 10% cross-linked, strongly acidic gel type, 2.0 meq/ml on PS-DVB
Brand:  Thermo Scientific Alfa Aesar 046575.A1
Diaion™ SK110(Na), ion exchange resin, 10% cross-linked, strongly acidic gel type, 2.0 meq/ml on PS-DVB
Code : NEW
Additional Details : Weight : 1.00000kg
Product Code. 15444055