missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Diagnostic Kit for TSH (MPQuanti™ only)

Product Code. 30280602 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
Thyroid Stimulating Hormone (TSH)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes unitSize
30280602 Thyroid Stimulating Hormone (TSH) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280602 Supplier MP Biomedicals Supplier No. 07TSH5025
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Diagnostic Kit for Thyroid Stimulating Hormone (TSH) is intended for in-vitro quantitative determination of Thyroid Stimulating Hormone (TSH) in human serum, and plasma.

  • Diagnostic Kit for TSH (Serum/Plasma) is based on fluorescence immunoassay technology.
  • Measurement of TSH is useful to aid in screening the adult population for primary hypothyroidism by medical professionals. It could also be used in screening neonates for hypothyroidism.
  • TSH Test Cassette (Serum/Plasma) detects Thyroid Stimulating Hormone through Immunochromatographic quantitative detection technology.
  • The sample moves through the strip from sample pad to absorbent pad by the chromatographic force.
  • If the test sample contains TSH, it attaches to the TSH antibody which is conjugated with fluorescent microspheres.
  • Then the complex will be captured by the capture antibody coated on the nitrocellulose membrane (Test line).
  • The concentration of TSH in the sample correlates linearly with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and product standard curve, the concentration of TSH in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer to show TSH concentration in the sample.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 0.1-100μIU/mL, R≥0.990
  • Test results should be interpreted at 15 minutes
  • Suitable for screening neonates and adults

Specifications

Detectable Analytes Thyroid Stimulating Hormone (TSH)
Detection Method Immunofluorescence
Sample Type Human serum, Human plasma
Type POC, Quantitative, TSH
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.