missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Diagnostic Kit for C-Reactive Protein (CRP) (MPQuanti™ only)

Product Code. 30280607 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 Tests/Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280607 25 Tests/Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280607 Supplier MP Biomedicals Supplier No. 07CRP5025
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Diagnostic kit for C-reactive Protein is intended for in-vitro quantitative determination of C-reactive protein (CRP) in human whole blood, serum, or plasma.

  • Diagnostic kit for C-reactive Protein (Whole Blood/Serum/plasma) is based on fluorescence immunoassay technology.
  • The sample moves through the strip from sample pad to absorbent pad. If the specimen contains CRP, it attaches to the fluorescent microspheres-conjugated anti-CRP antibodies.
  • Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
  • The concentration of CRP in the sample correlates linearly with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and standard curve, the concentration of CRP in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 5 -100mg/L, R≥0.990
  • Test results should be interpreted at 3 minutes

Specifications

Detectable Analytes C-Reactive Protein
Detection Method Immunofluorescence
Sample Type Human whole blood, Human serum, Human plasma
Type POC, CRP, Quantitative
Quantity 25 Tests/Kit
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.