missing translation for 'onlineSavingsMsg'
Learn More

Diacetyl, Extra Pure, SLR, Fisher Chemical

Product Code. 10050060 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10050060 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10050060 Supplier Fisher Chemical Supplier No. D/0250/05

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 431-03-8
Melting Point -4°C
Color Yellow
pH 3.2
Boiling Point 88°C
Packaging Amber glass bottle
Molecular Formula C4H6O2
MDL Number 8756
Quantity 100 mL
InChI Key QSJXEFYPDANLFS-UHFFFAOYSA-N
SMILES CC(=O)C(C)=O
IUPAC Name butane-2,3-dione
Molecular Weight (g/mol) 86.09
Formula Weight 86.09g/mol
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.