missing translation for 'onlineSavingsMsg'
Learn More

Diacetyl (6-Chloro 7-Piperazinyl 1-Ethyl-1,4-dihydro-4-oxo-3-quinolinecarboxy Borate), TRC

Product Code. 30387799
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387799 100 mg 100mg
30387498 50 mg 50mg
30370716 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387799 Supplier Toronto Research Chemicals Supplier No. TRCD679330100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diacetyl (6-Chloro 7-Piperazinyl 1-Ethyl-1,4-dihydro-4-oxo-3-quinolinecarboxy Borate)
Analyte or Component Names Diacetyl (6-Chloro 7-Piperazinyl 1-Ethyl-1,4-dihydro-4-oxo-3-quinolinecarboxy Borate)
Shipping Condition Room Temperature
Molecular Formula C20 H23 B Cl N3 O7
InChI Formula InChI=1S/C20H23BClN3O7/c1-4-24-11-15(20(29)32-21(30-12(2)26)31-13(3)27)19(28)14-9-16(22)18(10-17(14)24)25-7-5-23-6-8-25/h9-11,23H,4-8H2,1-3H3
Solution Type Neat
SMILES CCN1C=C(C(=O)OB(OC(=O)C)OC(=O)C)C(=O)c2cc(Cl)c(cc12)N3CCNCC3
IUPAC Name diacetyloxyboranyl 6-chloro-1-ethyl-4-oxo-7-piperazin-1-ylquinoline-3-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 463.676
Quantity 100 mg
Formula Weight 463.132
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.