missing translation for 'onlineSavingsMsg'
Learn More

(Diacetoxyiodo)benzene, TRC

Product Code. 17973555
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973555 100 g 100g
17945075 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17973555 Supplier Toronto Research Chemicals Supplier No. TRCD360500100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3240-34-4
Molecular Formula C10 H11 I O4
Molecular Weight (g/mol) 322.1
Synonym (Dihydroxyiodo)benzene, (Diacetoxyiodo)benzene, BAIB, Bis(acetato)phenyliodine, Bis(acetato-κO)phenyliodine, Diacetoxy(phenyl)iodine, Iodobenzene Diacetate, Iodophenyl Diacetate, Iodosobenzene Diacetate, Iodosylbenzene Diacetate, NSC 226375, NSC 23801, PIDA, Phenyliodine Diacetate, Phenyliodine(III) Diacetate, Phenyliodo Diacetate, Phenyliodonium Diacetate, Phenyliodoso Acetate, Phenyliodoso Diacetate, Phenyliodosyl Diacetate, [Bis(acetoxy)iodo]benzene, Iodosodiacetate Benzene;
IUPAC Name [acetoxy(phenyl)-λ3-iodanyl] acetate
SMILES CC(=O)OI(C1=CC=CC=C1)OC(=O)C

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H11IO4/c1-8(12)14-11(15-9(2)13)10-6-4-3-5-7-10/h3-7H,1-2H3
Formula Weight 321.9702
Chemical Name or Material (Diacetoxyiodo)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.