missing translation for 'onlineSavingsMsg'
Learn More

Diacetone acrylamide, 99%

Product Code. 11488476
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488476 50 g 50g
11498476 250 g 250g
11478476 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488476 Supplier Thermo Scientific Alfa Aesar Supplier No. A15940.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2873-97-4 | C9H15NO2 | 169.22 g/mol

Diacetone acrylamide is used in epoxy resin, acryl emulsion, water based paints, waste water treatment, photosensitive resins, waterborne coatings and gel electrophoresis. It is also used in the manufacture of permanent press fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diacetone acrylamide is used in epoxy resin, acryl emulsion, water based paints, waste water treatment, photosensitive resins, waterborne coatings and gel electrophoresis. It is also used in the manufacture of permanent press fabrics.

Solubility
Soluble in water, methanol, benzene and tetrahydrofuran.

Notes
Store in cool place. Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2873-97-4
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.22
MDL Number MFCD00008788
InChI Key OMNKZBIFPJNNIO-UHFFFAOYSA-N
Synonym diacetone acrylamide, diacetoneacrylamide, n-1,1-dimethyl-3-oxobutyl acrylamide, 2-propenamide, n-1,1-dimethyl-3-oxobutyl, n-2-methyl-4-oxopentan-2-yl acrylamide, n-2-2-methyl-4-oxopentyl acrylamide, acrylamide, n-1,1-dimethyl-3-oxobutyl, n-1,1-dimethyl-3-oxobutyl-2-propenamide, acrylamide, n,n-diacetonyl, ccris 5898
PubChem CID 17888
IUPAC Name N-(2-methyl-4-oxopentan-2-yl)prop-2-enamide
SMILES CC(=O)CC(C)(C)NC(=O)C=C

Specifications

Melting Point 54°C to 56°C
Flash Point >110°C (230°F)
Quantity 50 g
Beilstein 1928444
Sensitivity Hygroscopic
Merck Index 14,2963
Solubility Information Soluble in water,methanol,benzene and tetrahydrofuran.
Formula Weight 169.23
Percent Purity 99%
Chemical Name or Material Diacetone acrylamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.