missing translation for 'onlineSavingsMsg'
Learn More

Diacetin, 50%, tech. grade, remainder triacetin and monoacetin, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 406300050

Code : 29

Additional Details : CAS Number : 25395-31-7 Weight : 0.00500kg

 View more versions of this product

Product Code. 10685481

  • £15.90 / 5g
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diacetin
25395-31-7,102-76-1,26446-35-5
1.1875g/mL
141°C
45% min. (GC)
C7H12O5
(CH3COO)2C3H5OH
5 g
1.1875
Solubility in water: soluble
176.17
24 to 28% monoacetin, 41 to 47% diacetin, 22 to 28% triacetin (typically)
pract., 98%
Colorless
280.0°C
Authentic
Glass bottle
1.4400 to 1.4500
MFCD00008717
542 to 605mg KOH/g
15, 2964
176.17
50%
Liquid
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only