missing translation for 'onlineSavingsMsg'
Learn More

Diacetin, 50%, tech. grade, remainder triacetin and monoacetin

Product Code. 10157130 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10157130 500 g Glass bottle 500g
10685481 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10157130 Supplier Thermo Scientific Acros Supplier No. 406305000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25395-31-7 | C7H12O5 | 176.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diacetin
Name Note pract., 98%
CAS 25395-31-7, 102-76-1, 26446-35-5
Color Colorless
Density 1.1875g/mL
Boiling Point 280.0°C
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 45% min. (GC)
Packaging Glass bottle
Molecular Formula C7H12O5
Linear Formula (CH3COO)2C3H5OH
Refractive Index 1.4400 to 1.4500
MDL Number MFCD00008717
Quantity 500 g
Saponification Value 542 to 605mg KOH/g
Merck Index 15, 2964
Specific Gravity 1.1875
Solubility Information Solubility in water: soluble
Molecular Weight (g/mol) 176.17
Formula Weight 176.17
Percent Purity 50%
Concentration or Composition (by Analyte or Components) 24 to 28% monoacetin, 41 to 47% diacetin, 22 to 28% triacetin (typically)
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.