missing translation for 'onlineSavingsMsg'
Learn More

Diacetin, mixed isomers, tech. ca 50%, remainder triacetin and monoacetin

Product Code. p-7045990
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11353326

Brand: Thermo Scientific Alfa Aesar L12708.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25395-31-7 | C7H12O5

Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25395-31-7
Molecular Formula C7H12O5
MDL Number MFCD00008717
Synonym glyceryl diacetate, 1,2-diacetin, glycerol diacetate, 1,2,3-propanetriol, diacetate, 3-hydroxypropane-1,2-diyl diacetate, glycerol 1,2-diacetate, 1,2-diacylglycerol, 2,3-diacetin, 1,2-diacetylglycerol, acetin, di

Specifications

Density 1.19
Boiling Point 250°C to 280°C (decomposition)
Flash Point 141°C (285°F)
Refractive Index 1.444
Quantity 250 g
Beilstein 1706903
Sensitivity Hygroscopic
Merck Index 14,2961
Solubility Information Fully miscible in water.
Formula Weight 176.17
Percent Purity ∼50%
Grade Technical
Assay remainder triacetin and monoacetin
Chemical Name or Material Diacetin, mixed isomers
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.