missing translation for 'onlineSavingsMsg'
Learn More

Diacetin, mixed isomers, tech. ca 50%, remainder triacetin and monoacetin

Product Code. 11363326
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363326 1000 g 1kg
11353326 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363326 Supplier Thermo Scientific Alfa Aesar Supplier No. L12708.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25395-31-7 | C7H12O5

Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25395-31-7
Molecular Formula C7H12O5
MDL Number MFCD00008717
Synonym glyceryl diacetate, 1,2-diacetin, glycerol diacetate, 1,2,3-propanetriol, diacetate, 3-hydroxypropane-1,2-diyl diacetate, glycerol 1,2-diacetate, 1,2-diacylglycerol, 2,3-diacetin, 1,2-diacetylglycerol, acetin, di

Specifications

Density 1.19
Boiling Point 250°C to 280°C (decomposition)
Flash Point 141°C (285°F)
Refractive Index 1.444
Quantity 1000 g
Beilstein 1706903
Sensitivity Hygroscopic
Merck Index 14,2961
Solubility Information Fully miscible in water.
Formula Weight 176.17
Percent Purity ∼50%
Grade Technical
Assay remainder triacetin and monoacetin
Chemical Name or Material Diacetin, mixed isomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.