missing translation for 'onlineSavingsMsg'
Learn More

Diacerein, TRC

Product Code. 30356980
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356980 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356980 Supplier Toronto Research Chemicals Supplier No. TRCD30300050MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diacetyl Rhein (Diacerein)
CAS 13739-02-1
Molecular Formula C19 H12 O8
Quantity 50 mg
Synonym 4,5-Bis(acetyloxy)-9,10-dihydro-9,10-dioxo-2-anthracenecarboxylic acid, 9,10-Dihydro-4,5-dihydroxy-9,10-dioxo-2-anthroic acid diacetate, Rhein diacetate, 1,8-Diacetoxy-3-carboxyanthraquinone, 4,5-Diacetoxy-9,10-dioxoanthracene-2-carboxylic acid, 4,5-Diacetoxyanthraquinone-2-carboxylic acid, 4,5-Diacetylrhein, Artrodar, DAR, Diacerein, Diacerhein, Diacetylrhein, Fisiodar, Osteocerein, SF 277
InChI Formula InChI=1S/C19H12O8/c1-8(20)26-13-5-3-4-11-15(13)18(23)16-12(17(11)22)6-10(19(24)25)7-14(16)27-9(2)21/h3-7H,1-2H3,(H,24,25)
SMILES CC(=O)Oc1cccc2C(=O)c3cc(cc(OC(=O)C)c3C(=O)c12)C(=O)O
IUPAC Name 4,5-diacetyloxy-9,10-dioxoanthracene-2-carboxylic acid
Molecular Weight (g/mol) 368.29
Formula Weight 368.0532
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.