missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diacerein, TRC
Specifications
Specifications
| Chemical Name or Material | Diacetyl Rhein (Diacerein) |
| CAS | 13739-02-1 |
| Molecular Formula | C19 H12 O8 |
| Quantity | 500 mg |
| Synonym | 4,5-Bis(acetyloxy)-9,10-dihydro-9,10-dioxo-2-anthracenecarboxylic acid, 9,10-Dihydro-4,5-dihydroxy-9,10-dioxo-2-anthroic acid diacetate, Rhein diacetate, 1,8-Diacetoxy-3-carboxyanthraquinone, 4,5-Diacetoxy-9,10-dioxoanthracene-2-carboxylic acid, 4,5-Diacetoxyanthraquinone-2-carboxylic acid, 4,5-Diacetylrhein, Artrodar, DAR, Diacerein, Diacerhein, Diacetylrhein, Fisiodar, Osteocerein, SF 277 |
| InChI Formula | InChI=1S/C19H12O8/c1-8(20)26-13-5-3-4-11-15(13)18(23)16-12(17(11)22)6-10(19(24)25)7-14(16)27-9(2)21/h3-7H,1-2H3,(H,24,25) |
| SMILES | CC(=O)Oc1cccc2C(=O)c3cc(cc(OC(=O)C)c3C(=O)c12)C(=O)O |
| IUPAC Name | 4,5-diacetyloxy-9,10-dioxoanthracene-2-carboxylic acid |
| Molecular Weight (g/mol) | 368.29 |
| Formula Weight | 368.0532 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?