missing translation for 'onlineSavingsMsg'
Learn More

Diacerein, TRC

Product Code. 30354111
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30354111 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354111 Supplier Toronto Research Chemicals Supplier No. TRCD3030005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diacetyl Rhein (Diacerein)
CAS 13739-02-1
Molecular Formula C19 H12 O8
Quantity 5 g
Synonym 4,5-Bis(acetyloxy)-9,10-dihydro-9,10-dioxo-2-anthracenecarboxylic acid, 9,10-Dihydro-4,5-dihydroxy-9,10-dioxo-2-anthroic acid diacetate, Rhein diacetate, 1,8-Diacetoxy-3-carboxyanthraquinone, 4,5-Diacetoxy-9,10-dioxoanthracene-2-carboxylic acid, 4,5-Diacetoxyanthraquinone-2-carboxylic acid, 4,5-Diacetylrhein, Artrodar, DAR, Diacerein, Diacerhein, Diacetylrhein, Fisiodar, Osteocerein, SF 277
InChI Formula InChI=1S/C19H12O8/c1-8(20)26-13-5-3-4-11-15(13)18(23)16-12(17(11)22)6-10(19(24)25)7-14(16)27-9(2)21/h3-7H,1-2H3,(H,24,25)
SMILES CC(=O)Oc1cccc2C(=O)c3cc(cc(OC(=O)C)c3C(=O)c12)C(=O)O
IUPAC Name 4,5-diacetyloxy-9,10-dioxoanthracene-2-carboxylic acid
Molecular Weight (g/mol) 368.29
Formula Weight 368.0532
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.