missing translation for 'onlineSavingsMsg'
Learn More

Diacerein

Product Code. 15425949
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425949 5 g 5g
15415949 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425949 Supplier Thermo Scientific Alfa Aesar Supplier No. J63775.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Interleukin-1 inhibitor | CAS: 13739-02-1 | C19H12O8 | 368.30 g/mol

Interleukin-1 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13739-02-1
Molecular Formula C19H12O8
Molecular Weight (g/mol) 368.30
MDL Number MFCD00468030
InChI Key TYNLGDBUJLVSMA-UHFFFAOYSA-N
Synonym diacerein, diacetylrhein, diacerhein, artrodar, fisiodar, 1,8-diacetoxy-3-carboxyanthraquinone, 4,5-diacetylrhein, diacerin, diacerein inn, diacereine french
PubChem CID 26248
IUPAC Name 4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid
SMILES CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O

Specifications

Melting Point 217°C to 218°C
Color Yellow
Quantity 5 g
Merck Index 14,2960
Formula Weight 368.29
Physical Form Powder
Chemical Name or Material Diacerein

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.