missing translation for 'onlineSavingsMsg'
Learn More

Di(tetrahydropyran-4-yl)amine, 97%, Thermo Scientific™

Product Code. p-3881238
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10596134

Brand: Thermo Scientific Maybridge MO08504DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1080028-76-7
Molecular Formula C10H19NO2
Molecular Weight (g/mol) 185.267
MDL Number MFCD11841081
InChI Key WWPWWQFNGOWXOS-UHFFFAOYSA-N
Synonym bis 4-tetrahydropyranyl amine, n-oxan-4-yl oxan-4-amine, di tetrahydropyran-4-yl amine, bis tetrahydropyran-4-yl amine, n-tetrahydro-2h-pyran-4-yl tetrahydro-2h-pyran-4-amine, n-4-oxanyl-4-oxanamine, bis tetrahydro-2h-pyran-4-yl amine, di 2h-3,4,5,6-tetrahydropyran-4-yl amine
PubChem CID 33589627
IUPAC Name N-(oxan-4-yl)oxan-4-amine
SMILES C1COCCC1NC2CCOCC2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 185.27
Percent Purity 97%
Chemical Name or Material Di(tetrahydropyran-4-yl)amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.