missing translation for 'onlineSavingsMsg'
Learn More

Di(tetrahydropyran-4-yl)amine, 97%, Thermo Scientific™

Product Code. 10173134
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173134 5 g Amber Glass Bottle 5g
10596134 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10173134 Supplier Thermo Scientific Maybridge Supplier No. MO08504DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1080028-76-7
Molecular Formula C10H19NO2
Molecular Weight (g/mol) 185.267
MDL Number MFCD11841081
InChI Key WWPWWQFNGOWXOS-UHFFFAOYSA-N
Synonym bis 4-tetrahydropyranyl amine, n-oxan-4-yl oxan-4-amine, di tetrahydropyran-4-yl amine, bis tetrahydropyran-4-yl amine, n-tetrahydro-2h-pyran-4-yl tetrahydro-2h-pyran-4-amine, n-4-oxanyl-4-oxanamine, bis tetrahydro-2h-pyran-4-yl amine, di 2h-3,4,5,6-tetrahydropyran-4-yl amine
PubChem CID 33589627
IUPAC Name N-(oxan-4-yl)oxan-4-amine
SMILES C1COCCC1NC2CCOCC2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 185.27
Percent Purity 97%
Chemical Name or Material Di(tetrahydropyran-4-yl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.