missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-octyl Disulfide (>80%), TRC

Product Code. 30381609
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381609 250 mg -
30386119 2.5 g 2.5g
30387963 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381609 Supplier Toronto Research Chemicals Supplier No. TRCD482155250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29956-99-8
Molecular Formula C16H34S2
Molecular Weight (g/mol) 290.57
IUPAC Name 2,2,4-trimethyl-4-(2,4,4-trimethylpentan-2-yldisulfanyl)pentane
SMILES CC(C)(C)CC(C)(C)SSC(C)(C)CC(C)(C)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C16H34S2/c1-13(2,3)11-15(7,8)17-18-16(9,10)12-14(4,5)6/h11-12H2,1-10H3
Formula Weight 290.2102
Chemical Name or Material Di-tert-octyl Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.