missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyltin dichloride, 98%

Product Code. 11314567
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
This item is not returnable. View return policy

Product Code. 11314567

Brand: Thermo Scientific Alfa Aesar 039619.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19429-30-2 | C8H18Cl2Sn | 303.842 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19429-30-2
Molecular Formula C8H18Cl2Sn
Molecular Weight (g/mol) 303.842
MDL Number MFCD00009854
InChI Key PEGCFRJASNUIPX-UHFFFAOYSA-L
Synonym di-tert-butyldichlorostannane, di-tert-butyltin dichloride, di-t-butyltin dichloride, ditert-butyl dichloro stannane, acmc-1bpuo, di-tert-butylzinndichlorid
PubChem CID 88053
IUPAC Name ditert-butyl(dichloro)stannane
SMILES CC(C)(C)[Sn](C(C)(C)C)(Cl)Cl

Specifications

Melting Point 42°C to 43°C
Color White
Boiling Point 66°C (3 mmHg)
Flash Point >110°C (230°F)
Odor Acrid
Linear Formula [(CH3)3C]2SnCl2
Quantity 2 g
UN Number UN2928
Sensitivity Moisture sensitive
Formula Weight 303.83
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Di-tert-butyltin dichloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.