missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butylphosphine, 0.5M (10 wt.%) solution in hexanes, AcroSeal™

Product Code. 10712962 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10712962 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10712962 Supplier Thermo Scientific Acros Supplier No. 429011000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 819-19-2 | C8H19P | 146.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Di-tert-butylphosphine
CAS 819-19-2, 110-54-3
Color Colorless
Density 0.6850g/mL
Flash Point −22°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C8H19P
Linear Formula [(CH3)3C]2PH
MDL Number MFCD00134241
Quantity 100 mL
Specific Gravity 0.685
Synonym di-tert-butyl-phosphane, di-tert-butylphosphine, di-t-butylphosphine, phosphine, bis 1,1-dimethylethyl, phosphine, di-tert-butyl, di-tert-butylphosphane, bis tert-butyl phosphine, hp t-bu 2, tbu2ph, di-tert-butylphoshine
Solubility Information Solubility in water: insoluble
InChI Key CRHWEIDCXNDTMO-UHFFFAOYSA-N
SMILES CC(C)(C)PC(C)(C)C
IUPAC Name ditert-butylphosphane
Molecular Weight (g/mol) 146.21
PubChem CID 136643
Formula Weight 146.21
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-tert-butylphosphine, 10 wt.% solution in hexanes
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.