missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butylphosphine, 0.5M (10 wt.%) solution in hexanes, Thermo Scientific Chemicals

£42.00 - £126.90

Specifications

Chemical Name or Material Di-tert-butylphosphine
CAS 819-19-2, 110-54-3
Color Colorless
Density 0.6850g/mL
Flash Point −22°C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10391903
View Documents Promotion Details
Thermo Scientific
383020100
10 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10301913
View Documents Promotion Details
Thermo Scientific
383020500
50 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Di-tert-butylphosphine
Colorless
−22°C
C8H19P
MFCD00134241
di-tert-butyl-phosphane, di-tert-butylphosphine, di-t-butylphosphine, phosphine, bis 1,1-dimethylethyl, phosphine, di-tert-butyl, di-tert-butylphosphane, bis tert-butyl phosphine, hp t-bu 2, tbu2ph, di-tert-butylphoshine
CRHWEIDCXNDTMO-UHFFFAOYSA-N
ditert-butylphosphane
136643
Liquid
819-19-2, 110-54-3
0.6850g/mL
Glass bottle
[(CH3)3C]2PH
0.685
Solubility in water: insoluble.
CC(C)(C)PC(C)(C)C
146.21
146.21
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or re

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

Product Identifier
  • Di-tert-butylphosphine, 10 wt.% solution in hexanes

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only