missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl Peroxide, TRC

Product Code. 30375918
Change view
Click to view available options
Quantity:
1 mL
2 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30375918 10 mL
30381764 2 mL
30374912 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30375918 Supplier Toronto Research Chemicals Supplier No. TRCD49529310ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-05-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
IUPAC Name 2-tert-butylperoxy-2-methylpropane
SMILES CC(C)(C)OOC(C)(C)C

Specifications

Quantity 10 mL
InChI Formula InChI=1S/C8H18O2/c1-7(2,3)9-10-8(4,5)6/h1-6H3
Formula Weight 146.1307
Chemical Name or Material Di-tert-butyl Peroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.