missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl N,N-diisopropylphosphoramidite, 95%, Thermo Scientific Chemicals

Product Code. 10172814
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137348-86-8
Molecular Formula C14H32NO2P
Molecular Weight (g/mol) 277.39
MDL Number MFCD00153506
InChI Key YGFLCNPXEPDANQ-UHFFFAOYSA-N
Synonym di-tert-butyl diisopropylphosphoramidite, di-tert-butyl n,n-diisopropylphosphoramidite, di-t-butyl n,n-diisopropylphosphoramidite, di-tert-butoxy diisopropylamino phosphine, n-di-tert-butoxyphosphanyl-n-isopropyl-propan-2-amine, phosphoramidous acid, bis 1-methylethyl-, bis 1,1-dimethylethyl ester, bis tert-butoxy phosphino bis methylethyl amine, bis tert-butoxy phosphanyl diisopropylamine, acmc-1by13, di-tertbutyl diisopropyl-phosphoramidite
PubChem CID 853005
IUPAC Name N-[bis[(2-methylpropan-2-yl)oxy]phosphanyl]-N-propan-2-ylpropan-2-amine
SMILES CC(C)N(C(C)C)P(OC(C)(C)C)OC(C)(C)C

Specifications

Color Colorless
Density 0.8790g/mL
Boiling Point 85.0°C to 90.0°C (0.2 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4390 to 1.4410 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.879
Formula Weight 277.39
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Di-tert-butyl N, N-diisopropylphosphoramidite
Show More Show Less
compliance-icons
Product Identifier
  • Di-tert-butyl N,N-diisopropylphosphoramidite
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'