missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl malonate, 98+%, stab. with potassium carbonate

Product Code. 11465113
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465113 10 g 10g
11475113 50 g 50g
11485113 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465113 Supplier Thermo Scientific Alfa Aesar Supplier No. A12774.09
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 541-16-2 | C11H20O4 | 216.277 g/mol

Di-tert-butyl malonate, is used as a pharmaceutical intermediate. it can be used to produce Diazomalonsaeure-di-tert-butylester at the ambient temperature. It will need reagents cesium carbonate, p-toluenesulfonyl azide and solvent tetrahydrofuran with the reaction time of 1 hour. T

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butyl malonate, is used as a pharmaceutical intermediate. it can be used to produce Diazomalonsaeure-di-tert-butylester at the ambient temperature. It will need reagents cesium carbonate, p-toluenesulfonyl azide and solvent tetrahydrofuran with the reaction time of 1 hour. T

Solubility
It is hardly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-16-2
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00008810
InChI Key CLPHAYNBNTVRDI-UHFFFAOYSA-N
Synonym di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate
PubChem CID 68324
IUPAC Name ditert-butyl propanedioate
SMILES CC(C)(C)OC(=O)CC(=O)OC(C)(C)C

Specifications

Melting Point -6°C
Density 0.965
Boiling Point 65°C to 67°C (1 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.419
Quantity 10 g
Beilstein 1781766
Merck Index 14,3034
Solubility Information It is hardly soluble in water.
Formula Weight 216.28
Percent Purity ≥98%
Chemical Name or Material Di-tert-butyl malonate, stabilized with potassium carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.