missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl iminodicarboxylate, 98%

Product Code. 11349176
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349176 25 g 25g
11329176 1 g 1g
11339176 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349176 Supplier Thermo Scientific Alfa Aesar Supplier No. L20325.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51779-32-9 | C10H19NO4 | 217.27 g/mol

Di-tert-butyl-iminodicarboxylate may be used in the preparation of trans-diamino-2-butene and cis-1,4-diamino-2-butene; di-tert-butyl N-3-butenyliminodicarboxylate; N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate & C1-C20 and C21-C40 fragments of tetrafibricin. Modified Gabriel Reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butyl-iminodicarboxylate may be used in the preparation of trans-diamino-2-butene and cis-1,4-diamino-2-butene; di-tert-butyl N-3-butenyliminodicarboxylate; N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate & C1-C20 and C21-C40 fragments of tetrafibricin. Modified Gabriel Reagent.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51779-32-9
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00043309
InChI Key XCAQIUOFDMREBA-UHFFFAOYSA-N
Synonym di-tert-butyl iminodicarboxylate, di-t-butyl iminodicarboxylate, di-tert-butyl imidodicarbonate, n-boc-tert-butylcarbamate, tert-butyl iminodicarboxylate, di-tert-butyl iminodicarbonate, di-tert-butyl-iminodicarboxylate, iminodicarboxylic acid di-tert-butyl ester, tert-butyl n-tert-butoxy carbonyl carbamate, di tert-butyl imidodicarbonate
PubChem CID 279800
IUPAC Name tert-butyl N-[(2-methylpropan-2-yl)oxycarbonyl]carbamate
SMILES CC(C)(C)OC(=O)NC(=O)OC(C)(C)C

Specifications

Melting Point 114°C to 119°C
Beilstein 1911172
Solubility Information Insoluble in water.
Quantity 25 g
Formula Weight 217.27
Sensitivity Light sensitive
Percent Purity 98%
Chemical Name or Material Di-tert-butyl iminodicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.