missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl hydrazodicarboxylate, 98+%

Product Code. 11319973
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319973 5 g 5g
11329973 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319973 Supplier Thermo Scientific Alfa Aesar Supplier No. L00280.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16466-61-8 | C10H20N2O4 | 232.28 g/mol

Di-tert-butyl hydrazodicarboxylate, is used as starting reagent during the four-step synthesis of the aminobicyclopyrazolone hydrochloride. It was used as reagent during Pd-catalyzed amination reactions of halo-pyridine substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butyl hydrazodicarboxylate, is used as starting reagent during the four-step synthesis of the aminobicyclopyrazolone hydrochloride. It was used as reagent during Pd-catalyzed amination reactions of halo-pyridine substrates.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16466-61-8
Molecular Formula C10H20N2O4
Molecular Weight (g/mol) 232.28
MDL Number MFCD00015000
InChI Key TYSZETYVESRFNT-UHFFFAOYSA-N
Synonym di-tert-butyl hydrazodicarboxylate, di-tert-butyl hydrazine-1,2-dicarboxylate, di-tert-butyl hydrazodiformate, di-tert-butyl bicarbamate, n,n'-di-boc-hydrazine, n,n'-bis tert-butoxycarbonyl hydrazine, di-tert-butyl-1,2-hydrazodicarboxylate, 1,2-hydrazinedicarboxylic acid, bis 1,1-dimethylethyl ester, tert-butyl n-2-methylpropan-2-yl oxycarbonylamino carbamate, n'-tert-butoxy carbonyl tert-butoxy carbohydrazide
PubChem CID 85431
IUPAC Name tert-butyl N-[(2-methylpropan-2-yl)oxycarbonylamino]carbamate
SMILES CC(C)(C)OC(=O)NNC(=O)OC(C)(C)C

Specifications

Melting Point 123°C to 126°C
Quantity 5 g
Beilstein 1794023
Solubility Information Insoluble in water.
Formula Weight 232.28
Percent Purity ≥98%
Chemical Name or Material Di-tert-butyl hydrazodicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.