missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl Glutamate Hydrochloride, TRC

Product Code. 30385239
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30385239

Brand: Toronto Research Chemicals TRCD49373510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Di-tert-butyl Glutamate Hydrochloride
CAS 32677-01-3
Molecular Formula C13H26ClNO4
Quantity 10 g
Synonym L-Glutamic Acid 1,5-Bis(1,1-dimethylethyl) Ester Hydrochloride, L-Glutamic Acid Bis(1,1-dimethylethyl) Ester Hydrochloride, Di-tert-butyl (S)-2-Aminopentanedioate Hydrochloride, Di-tert-butyl L-Glutamate Hydrochloride, Di-tert-butyl Glutamate Hydrochloride, Glutamic Acid Di-tert-butyl Ester Hydrochloride, L-Glutamic Acid Di-tert-butyl Ester Hydrochloride, NSC 94681
InChI Formula InChI=1S/C13H25NO4.ClH/c1-12(2,3)17-10(15)8-7-9(14)11(16)18-13(4,5)6;/h9H,7-8,14H2,1-6H3;1H/t9-;/m0./s1
SMILES N[C@@H](CCC(OC(C)(C)C)=O)C(OC(C)(C)C)=O.Cl
IUPAC Name di-tert-butyl L-glutamate hydrochloride
Molecular Weight (g/mol) 295.8
Formula Weight 295.16
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.