missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-tert-butyl dicarbonate, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Reagent for the introduction of the Boc protecting group.Di-tert-butyl dicarbonate is a reagent used for the introduction of BOC protecting group. It plays an important role in the preparation of 6-acetyl-1,2,3,4-tetrahydropyridine by reacting with 2-piperidone. It serves as a protecting group used in solid phase peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reagent for the introduction of the Boc protecting group.Di-tert-butyl dicarbonate is a reagent used for the introduction of BOC protecting group. It plays an important role in the preparation of 6-acetyl-1,2,3,4-tetrahydropyridine by reacting with 2-piperidone. It serves as a protecting group used in solid phase peptide synthesis.
Solubility
Miscible with decalin, toluene, carbon tetrachloride, tetrahydrofuran, dioxane, alcohols, acetone, acetonitrile and dimethylformamide. Immiscible with water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Moisture and light sensitive. Incompatible with oxidizing agents, reducing agents, bases and acids.
Reagent for the introduction of the Boc protecting group.Di-tert-butyl dicarbonate is a reagent used for the introduction of BOC protecting group. It plays an important role in the preparation of 6-acetyl-1,2,3,4-tetrahydropyridine by reacting with 2-piperidone. It serves as a protecting group used in solid phase peptide synthesis.
Solubility
Miscible with decalin, toluene, carbon tetrachloride, tetrahydrofuran, dioxane, alcohols, acetone, acetonitrile and dimethylformamide. Immiscible with water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Moisture and light sensitive. Incompatible with oxidizing agents, reducing agents, bases and acids.
Specifications
Melting Point | 21°C to 24°C |
Density | 0.950 |
Boiling Point | 56°C to 57°C (0.5 mmHg) |
Flash Point | 37°C (99°F) |
Assay Percent Range | ≥97% |
Refractive Index | 1.409 |
Quantity | 100 g |
UN Number | 2930 |
Beilstein | 1911173 |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'