missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl ChloromethylPhosphate (stabilized with K2CO3), TRC

Product Code. 30386898
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386898 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386898 Supplier Toronto Research Chemicals Supplier No. TRCD428028500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Di-tert-butyl Chloromethyl Phosphate
CAS 229625-50-7
Molecular Formula C9 H20 Cl O4 P
Quantity 500 mg
Synonym Solution, contains solid potassium carbonate for buffering purposes, GSK1053539A, Fostemsavir Tromethamine related (GSK3684934A (BMS-663068-03)), Phosphoric acid, chloromethyl bis(1,1-dimethylethyl) ester, Chloromethyl di-tert-butyl phosphate, Di-tert-butyl chloromethyl phosphate, Phosphoric acid di(tert-butyl) chloromethyl ester, Phosphoric acid ditert-butyl ester chloromethyl ester, BMS-371000
InChI Formula InChI=1S/C9H20ClO4P/c1-8(2,3)13-15(11,12-7-10)14-9(4,5)6/h7H2,1-6H3
SMILES CC(C)(C)OP(=O)(OCCl)OC(C)(C)C
IUPAC Name ditert-butyl chloromethyl phosphate
Molecular Weight (g/mol) 258.6795
Formula Weight 258.0788
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.