missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Sodium Hydrogen Citrate Sesquihydrate, 99-101%, Extra Pure, SLR, Fisher Chemical
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Specifications
Melting Point | 149°C |
CAS | 6142-05-8 |
Color | White |
pH | 4.9 to 5.2 |
Physical Form | Powder/Solid |
Quantity | 500 g |
Molecular Formula | C3H5ClN2S |
MDL Number | 150445 |
Synonym | 2-aminothiazole hydrochloride, thiazol-2-amine hydrochloride, thiazol-2-ylammonium chloride, 2-thiazolamine, monohydrochloride, aminothiazole hydrochloride, 1,3-thiazol-2-aminium chloride, 1,3-thiazol-2-amine hydrochloride, 1,3-thiazole-2-ylamine, chloride, ksc493o5f, thiazol-2-ylamine hydrochloride |
InChI Key | WDJXDOCQCALXMV-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'