missing translation for 'onlineSavingsMsg'
Learn More

Di-sec-butoxyaluminoxytriethoxysilane

Product Code. 11399577
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399577 50 g 50g
11309587 10 g 10g
11389577 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399577 Supplier Thermo Scientific Alfa Aesar Supplier No. 041210.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68959-06-8 | C14H33AlO6Si | 352.48 g/mol

Di-sec-butoxyaluminoxytriethoxysilane is used as Sol-gel intermediate for aluminum silicates. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68959-06-8
Molecular Formula C14H33AlO6Si
Molecular Weight (g/mol) 352.48
MDL Number MFCD00053771
InChI Key MTHHTRVJDXYJHE-UHFFFAOYNA-N
Synonym di-sec-butoxyaluminoxytriethoxysilane, bis butan-2-olato triethyl orthosilicato-o' aluminum, aluminum, bis 2-butanolato triethyl orthosilicato-.kappa.o', aluminum, bis 2-butanolato triethyl orthosilicato-kappao', di-s-butoxyaluminoxy-triethoxysilane, di-sec-butoxy triethoxysilyl oxy aluminum, bis sec-butoxy alumanyl triethyl silicate, aluminum,bis 2-butanolato triethyl orthosilicato-ko', diethoxysiloxane-s-butylaluminate copolymer di-s-butoxyaluminoxytriethoxysilane
PubChem CID 16684138
IUPAC Name di(butan-2-yloxy)alumanyl triethyl silicate
SMILES CCO[Si](OCC)(OCC)O[Al](OC(C)CC)OC(C)CC

Specifications

Boiling Point >200°C
Color Colorless
Physical Form Liquid
Quantity 50 g
UN Number UN1993
Flash Point 46°C (115°F)
Solubility Information Hydrolyzes with water.
Formula Weight 352.48
Sensitivity Moisture sensitive
Density 1.00 g/mL
Chemical Name or Material Di-sec-butoxyaluminoxytriethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.