Learn More
Di-sec-butoxyaluminoxytriethoxysilane
Description
Di-sec-butoxyaluminoxytriethoxysilane is used as Sol-gel intermediate for aluminum silicates. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | >200°C |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 250 g |
| UN Number | UN1993 |
| Flash Point | 46°C (115°F) |
| Solubility Information | Hydrolyzes with water. |
| Formula Weight | 352.48 |
| Sensitivity | Moisture sensitive |
| Density | 1.00 g/mL |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.