missing translation for 'onlineSavingsMsg'
Learn More

Dipotassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10734191 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10734191 500 g 500g
10030340 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10734191 Supplier Fisher Chemical Supplier No. P/5250/53
  • ONLINE EXCLUSIVE PRICE
    £697.00
    £136.00 / 500g
    Save £561.00
    80% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dipotassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 1kg, CAS: 16788-57-1, White, Molecular Formula: H7K2O7P, Molecular Weight: 228.219, Packaging: Plastic powder jar, pH: 8.5 to 9.6

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
IUPAC Name dipotassium;hydrogen phosphate;trihydrate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O

Specifications

Color White
pH 8.5 to 9.6
Physical Form Solid
Quantity 500 g
Packaging Plastic powder jar
Formula Weight 228.23g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.