missing translation for 'onlineSavingsMsg'
Learn More

(-)-Di-p-toluoyl-L-tartaric acid, 97%

Product Code. 10409030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10409030 25 g Glass bottle 25g
10580741 10 g Glass bottle 10g
10686734 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10409030 Supplier Thermo Scientific Acros Supplier No. 117570250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 32634-66-5 | C20H18O8 | 386.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32634-66-5
Molecular Formula C20H18O8
Molecular Weight (g/mol) 386.36
MDL Number MFCD00064440
InChI Key CMIBUZBMZCBCAT-IYBDPMFKSA-N
Synonym 2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid, di-p-toluoyl-l-tartaric acid, l-dtta, --di-p-toluoyl-l-tartaric acid, --di-1,4-o-toluoyl-l-tartaric acid, --o,o'-di-p-toluoyl-l-tartaric acid, l---di-p-toluoyltartaric acid, 2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid, butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r
PubChem CID 12297760
IUPAC Name (2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate
SMILES CC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 164°C to 170°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula [CH3C6H4CO2CH(CO2H)-]2
Quantity 25 g
Specific Rotation Condition −142° (20°C c=10,C2H5OH)
Specific Rotation -142
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone and ethanol
Formula Weight 386.36
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (-)-Di-p-toluoyl-L-tartaric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.